Mal-PEG6-NHS ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2,5-dioxopyrrolidin-1-yl 1-(2,5-dioxo-2H-pyrrol-1(5H)-yl)-3,6,9,12,15,18-hexaoxahenicosan-21-oate

A384375-1G A384375-250MG
77613-140EA 540.75 USD
77613-140 77686-069
Mal-PEG6-NHS ester ≥95%
Mal-PEG6-NHS ester

Precaución: for Research Use Only

Formula: C₂₃H₃₄N₂O₁₂
MDL Number: MFCD20926374
CAS Number: 1599472-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR