4-Bromo-2,3-difluoroiodobenzene ≥97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,3-difluoro-4-iodobenzene

A393118-1G A393118-5G
77684-124EA 66 USD
77684-124 77684-125
4-Bromo-2,3-difluoroiodobenzene ≥97%
4-Bromo-2,3-difluoroiodobenzene

Precaución: For research use only.

Formula: C₆H₂BrF₂I
MW: 318.885 g/mol
MDL Number: MFCD09878200
CAS Number: 207989-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR