L(-)-Xylose ≥97%

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4S)-2,3,4,5-Tetrahydroxypentanal, L-(-)-Xylose

A384206-100G A384206-25G A384206-500G A384206-5G A384206-10G
77601-686EA 85.08 USD
77601-686 77601-688 77601-690 77601-692 77668-788
L(-)-Xylose ≥97%
L(-)-Xylose

Precaución: for Research Use Only

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
MDL Number: MFCD00151096
CAS Number: 609-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR