α,α'-Dichloro-o-xylene ≥99%

Supplier: Ambeed
Danger

Synonyms: o-Xylylene Dichloride, o-Xylylene chloride, α,α′-Dichloro-o-xylene, α,α'-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, Alpha,Alpha’-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, o-Xylene dichloride

A383843-25G A383843-100G
77572-140EA 13.39 USD
77572-140 77626-752
α,α'-Dichloro-o-xylene ≥99%
α,α'-Dichloro-o-xylene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂
MW: 175.06 g/mol
MDL Number: MFCD00000903
CAS Number: 612-12-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR