1,3-Dithiane-2-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A383796-1G A383796-5G
77594-150EA 254.2 USD
77594-150 77746-772
1,3-Dithiane-2-carboxylic acid ≥98%
1,3-Dithiane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₈O₂S₂
MDL Number: MFCD07368267
CAS Number: 20461-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR