2,4,6-Tribromo-m-cresol 97+%

Supplier: Ambeed
Warning

A383350-5G A383350-1G A383350-25G A383350-100G
77598-464EA 20.5 USD
77598-464 77626-748 77626-750 77878-105
2,4,6-Tribromo-m-cresol 97+%
2,4,6-Tribromo-m-cresol

Precaución: for Research Use Only

Formula: C₇H₅Br₃O
MDL Number: MFCD00019976
CAS Number: 4619-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR