(R)-(+)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: (R)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl

A383256-1G A383256-250MG A383256-5G
77684-105EA 47 USD
77684-105 77684-106 77684-107
(R)-(+)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98%, Ambeed.Inc
(R)-(+)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl

Precaución: For research use only.

Formula: C₂₄H₂₀Br₂O₄
MW: 532.23 g/mol
MDL Number: MFCD03788936
CAS Number: 179866-74-1
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR