AZ-505 ditrifluoroacetate ≥98%

Supplier: Ambeed
Warning

Synonyms: N-cyclohexyl-3-[2-(3,4-dichlorophenyl)ethylamino]-N-[2-[2-(5-hydroxy-3-oxo-4H-1,4-benzoxazin-8-yl)ethylamino]ethyl]propenamide bis(2,2,2-trifluoroacetate), AZ505 ditrifluoroacetate, N-Cyclohexyl-3-((3,4-dichlorophenethyl)amino)-N-(2-((2-(5-hydroxy-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazin-8-yl)ethyl)amino)ethyl)propanamide bis(2,2,2-trifluoroacetate)

A391113-5MG A391113-1MG
77746-783EA 214 USD
77746-783 77746-782
AZ-505 ditrifluoroacetate ≥98%
AZ-505 ditrifluoroacetate

Precaución: for Research Use Only

Formula: C₃₃H₄₀Cl₂F₆N₄O₈
MW: 805.59 g/mol
MDL Number: MFCD28137733
CAS Number: 1035227-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR