8-Chloro-1-octanol ≥95%

Supplier: Ambeed
Warning

Synonyms: 8-Chloro-1-n-octanol

A382154-1G A382154-5G
77601-682EA 9.23 USD
77601-682 77601-684
8-Chloro-1-octanol ≥95%
8-Chloro-1-octanol

Precaución: for Research Use Only

Formula: C₈H₁₇ClO
MW: 164.68 g/mol
MDL Number: MFCD00040005
CAS Number: 23144-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR