3-(1-((3,3-Diphenylpropyl)(methyl)amino)-2-methylpropan-2-yl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate ≥98%

Supplier: Ambeed
Danger

Synonyms: Lercanidipine

A381766-100MG A381766-10MG A381766-5MG
77668-766EA 491 USD
77668-766 77668-768 77668-770
3-(1-((3,3-Diphenylpropyl)(methyl)amino)-2-methylpropan-2-yl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate ≥98%
3-(1-((3,3-Diphenylpropyl)(methyl)amino)-2-methylpropan-2-yl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₃₆H₄₁N₃O₆
MW: 611.73 g/mol
MDL Number: MFCD00867978
CAS Number: 100427-26-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR