5-Phenethylbenzene-1,3-diol ≥98%

Supplier: Ambeed
Warning

Synonyms: Dihydropinosylvin

A390006-250MG A390006-1G
77613-178EA 46.35 USD
77613-178 77672-940
5-Phenethylbenzene-1,3-diol ≥98%
5-Phenethylbenzene-1,3-diol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
MDL Number: MFCD16294420
CAS Number: 14531-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR