6-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one ≥98%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-4-methyl-2H-benzo[b][1,4]oxazin-3(4H)-one

A389925-250MG A389925-1G
77626-776EA 33.99 USD
77626-776 77878-124
6-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one ≥98%
6-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₂
MW: 178.19 g/mol
MDL Number: MFCD01834021
CAS Number: 103361-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR