(5-Bromo-2-methoxypyridin-3-yl)methanol ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-methoxy-3-pyridinemethanol

A380911-1G A380911-250MG
77900-891EA 40 USD
77900-891 77900-751
(5-Bromo-2-methoxypyridin-3-yl)methanol ≥98%
(5-Bromo-2-methoxypyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂
MDL Number: MFCD11840327
CAS Number: 351410-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR