6-Methoxyquinolin-2(1H)-one 95+%

Supplier: Ambeed
Warning

A380770-250MG A380770-1G
77901-820EA 143 USD
77901-820 77901-884
6-Methoxyquinolin-2(1H)-one 95+%
6-Methoxyquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD08688617
CAS Number: 13676-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR