Hydroxy-PEG5-CH2CO2tBu ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 17-hydroxy-3,6,9,12,15-pentaoxaheptadecan-1-oate, Hydroxy-PEG4-O-Boc
A380712-250MG
A380712-5G
A380712-100MG
A380712-25G
77748-764EA
47
USD
77748-764
77748-766
77748-763
77748-765
Hydroxy-PEG5-CH2CO2tBu ≥98%
Hydroxy-PEG5-CH2CO2tBu
Precaución: for Research Use Only
Formula:
C₁₆H₃₂O₈ MW: 352.42 g/mol |
MDL Number:
MFCD27977509 CAS Number: 1807530-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...