5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile ≥98%
Supplier: Ambeed
|
Warning
|
A380603-100MG
A380603-250MG
77884-011EA
128
USD
77884-011
77884-012
5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile ≥98%
Precaución: for Research Use Only
Formula:
C₈H₄BrN₃ |
MDL Number:
MFCD11046239 CAS Number: 799270-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...