Tetrafluorosuccinic acid ≥98%

Supplier: Ambeed
Danger

Synonyms: 2,2,3,3-Tetrafluorosuccinic acid, Tetrafluoro-1,4-butanedioic acid

A389066-1G A389066-25G
77872-609EA 20 USD
77872-609 77872-610
Tetrafluorosuccinic acid ≥98%
Tetrafluorosuccinic acid

Precaución: for Research Use Only

Formula: C₄H₂F₄O₄
MDL Number: MFCD00042430
CAS Number: 377-38-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR