trans-3,3-Dimethyl-1-butenylboronic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 2-tert-Butyl-trans-vinylboronic acid, (E)-(3,3-Dimethylbut-1-en-1-yl)boronic acid

A380571-250MG A380571-1G A380571-5G
77684-097EA 34 USD
77684-097 77892-784 77892-785
trans-3,3-Dimethyl-1-butenylboronic acid ≥98%, Ambeed.Inc
trans-3,3-Dimethyl-1-butenylboronic acid

Precaución: For research use only.

Formula: C₆H₁₃BO₂
MDL Number: MFCD01074624
CAS Number: 86595-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR