4-Chloro-7-fluoro-2-methylquinazoline ≥98%

Supplier: Ambeed
Warning

A388775-1G A388775-250MG
77572-172EA 33 USD
77572-172 77872-608
4-Chloro-7-fluoro-2-methylquinazoline ≥98%
4-Chloro-7-fluoro-2-methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₆ClFN₂
MDL Number: MFCD12024899
CAS Number: 1206694-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR