2-Methoxy-3-methylpyridine-5-boronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A380420-100MG A380420-1G A380420-250MG
77872-594EA 51 USD
77872-594 77872-595 77872-596
2-Methoxy-3-methylpyridine-5-boronic acid pinacol ester ≥95%
2-Methoxy-3-methylpyridine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₃
MDL Number: MFCD12923400
CAS Number: 1083168-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR