4-(Hydroxymethyl)-3-methoxybenzoic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A380312-1G A380312-250MG
77684-096EA 263 USD
77684-096 77892-782
4-(Hydroxymethyl)-3-methoxybenzoic acid ≥98%, Ambeed.Inc
4-(Hydroxymethyl)-3-methoxybenzoic acid

Precaución: For research use only.

Formula: C₉H₁₀O₄
MDL Number: MFCD15202211
CAS Number: 955117-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR