(3,5-Dibromophenyl)methanamine ≥95%

Supplier: Ambeed
Warning

A388602-5G A388602-1G A388602-250MG
77613-170EA 132.78 USD
77613-170 77892-798 77892-799
(3,5-Dibromophenyl)methanamine ≥95%
(3,5-Dibromophenyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₇Br₂N
MDL Number: MFCD03410983
CAS Number: 771580-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR