6-ECDCA ≥98%

Supplier: Ambeed
Warning

A387805-250MG A387805-5G A387805-1G A387805-100MG A387805-50MG A387805-10G A387805-25G
77613-162EA 22.66 USD
77613-162 77613-164 77613-160 77672-922 77672-924 77884-026 77884-027
6-ECDCA ≥98%
6-ECDCA

Precaución: for Research Use Only

Formula: C₂₆H₄₄O₄
MDL Number: MFCD16621104
CAS Number: 459789-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR