7-Bromo-5-fluoro-1H-indazole ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A387763-1G A387763-100MG A387763-250MG A387763-25G
77684-118EA 176 USD
77684-118 77886-025 77886-026 77886-027
7-Bromo-5-fluoro-1H-indazole ≥95%, Ambeed.Inc
7-Bromo-5-fluoro-1H-indazole

Precaución: For research use only.

Formula: C₇H₄BrFN₂
MDL Number: MFCD21602945
CAS Number: 1100214-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR