5-Bromo-2-fluoro-3-methylbenzeneboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (5-Bromo-2-fluoro-3-methylphenyl)boronic acid, 5-Bromo-2-fluoro-3-methylphenylboronic acid

A377486-1G A377486-25G A377486-5G
77672-870EA 22 USD
77672-870 77882-014 77882-015
5-Bromo-2-fluoro-3-methylbenzeneboronic acid ≥98%
5-Bromo-2-fluoro-3-methylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₂
MDL Number: MFCD09800868
CAS Number: 957120-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR