2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16,19,22,25,28,31,34-decaoxa-4-azaheptatriacontan-37-oate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Mal-amido-PEG10-C2-NHS ester
A376465-100MG
A376465-250MG
A376465-1G
77616-648EA
93.73
USD
77616-648
77616-650
77686-055
2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16,19,22,25,28,31,34-decaoxa-4-azaheptatriacontan-37-oate ≥98%
2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16,19,22,25,28,31,34-decaoxa-4-azaheptatriacontan-37-oate
Precaución: for Research Use Only
Formula:
C₃₄H₅₅N₃O₁₇ MW: 777.81 g/mol |
MDL Number:
MFCD20226950 CAS Number: 1137109-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...