Evobrutinib ≥98%

Supplier: Ambeed
Warning

Synonyms: MSC-2364447C, 1-(4-(((6-amino-5-(4-phenoxyphenyl)pyrimidin-4-yl)amino)methyl)piperidin-1-yl)prop-2-en-1-one, M-2951

A376209-1MG A376209-5MG A376209-10MG
77748-753EA 26 USD
77748-753 77748-754 77748-752
Evobrutinib ≥98%
Evobrutinib

Precaución: for Research Use Only

Formula: C₂₅H₂₇N₅O₂
MW: 429.52 g/mol
MDL Number: MFCD30502975
CAS Number: 1415823-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR