(S)-1-[3-(Trifluoromethyl)phenyl]ethanol ≥95%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(3-(Trifluoromethyl)phenyl)ethanol

A377992-100MG A377992-250MG
77886-000EA 18 USD
77886-000 77886-001
(S)-1-[3-(Trifluoromethyl)phenyl]ethanol ≥95%
(S)-1-[3-(Trifluoromethyl)phenyl]ethanol

Precaución: for Research Use Only

Formula: C₉H₉F₃O
MDL Number: MFCD03093004
CAS Number: 96789-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR