Bromoacetamido-PEG4-Acid ≥97%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-2-oxo-6,9,12,15-tetraoxa-3-azaoctadecan-18-oic acid

A369162-100MG A369162-250MG A369162-1G
77616-622EA 20.6 USD
77616-622 77616-624 77748-739
Bromoacetamido-PEG4-Acid ≥97%
Bromoacetamido-PEG4-Acid

Precaución: for Research Use Only

Formula: C₁₃H₂₄BrNO₇
MW: 386.236 g/mol
MDL Number: MFCD28950779
CAS Number: 1807518-67-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR