PH-797804 ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-(2,4-difluorobenzyloxy)-3-bromo-6-methyl-2-oxopyridin-1(2H)-yl)-N,4-dimethylbenzamide, 3-(3-Bromo-4-((2,4-difluorobenzyl)oxy)-6-methyl-2-oxopyridin-1(2H)-yl)-N,4-dimethylbenzamide, PH797804, PH 797804, 3-[3-bromo-4-[(2,4-difluorophenyl)methoxy]-6-methyl-2-oxopyridin-1-yl]-N,4-dimethylbenzamide, PH-797804

A369134-10MG A369134-1MG A369134-5MG
77613-088EA 177.16 USD
77613-088 77613-090 77613-092
PH-797804 ≥98%
PH-797804

Precaución: for Research Use Only

Formula: C₂₂H₁₉BrF₂N₂O₃
MW: 477.3 g/mol
MDL Number: MFCD18251426
CAS Number: 586379-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR