cis-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane ≥95%

Supplier: Ambeed
Warning

A375932-250MG A375932-100MG
77900-953EA 57 USD
77900-953 77901-274
cis-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane ≥95%
cis-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane

Precaución: for Research Use Only

Formula: C₁₃H₂₅NO₃
MDL Number: MFCD03844625
CAS Number: 1069120-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR