tert-Butyl 7-oxo-2-azaspiro[4.4]nonane-2-carboxylate ≥97%

Supplier: Ambeed
Warning

A375861-250MG A375861-1G
77686-051EA 261 USD
77686-051 77892-775
tert-Butyl 7-oxo-2-azaspiro[4.4]nonane-2-carboxylate ≥97%
tert-Butyl 7-oxo-2-azaspiro[4.4]nonane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₃
MW: 239.31 g/mol
MDL Number: MFCD23106459
CAS Number: 1319716-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR