1-tert-Butyl 3-methyl 5-oxopiperidine-1,3-dicarboxylate ≥98%

Supplier: Ambeed
Warning

A369118-1G A369118-250MG
77892-756EA 239 USD
77892-756 77892-757
1-tert-Butyl 3-methyl 5-oxopiperidine-1,3-dicarboxylate ≥98%
1-tert-Butyl 3-methyl 5-oxopiperidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₅
MDL Number: MFCD18909956
CAS Number: 1303974-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR