Abz-Gly-OH·HCl ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Abz-Gly-OH·HCl, 2-(2-Aminobenzamido)acetic acid hydrochloride
A375849-1G
A375849-250MG
77878-093EA
91
USD
77878-093
77878-094
Abz-Gly-OH·HCl ≥97%
Abz-Gly-OH·HCl
Precaución: for Research Use Only
Formula:
C₉H₁₁ClN₂O₃ MW: 230.648 g/mol |
MDL Number:
MFCD00237464 CAS Number: 256657-23-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...