5-Bromopyridine-2-ethanol ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A369080-5G A369080-1G
77684-072EA 260 USD
77684-072 77885-973
5-Bromopyridine-2-ethanol ≥98%, Ambeed.Inc
5-Bromopyridine-2-ethanol

Precaución: For research use only.

Formula: C₇H₈BrNO
MDL Number: MFCD09999981
CAS Number: 1206968-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR