Fexaramine ≥98%

Supplier: Ambeed
Warning

A375623-100MG A375623-50MG
77686-050EA 374 USD
77686-050 77900-537
Fexaramine ≥98%
Fexaramine

Precaución: for Research Use Only

Formula: C₃₂H₃₆N₂O₃
MDL Number: MFCD09971007
CAS Number: 574013-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR