Ethyl 6-oxohexanoate ≥95%

Supplier: Ambeed
Warning

A368741-5G A368741-1G
77748-737EA 143 USD
77748-737 77748-736
Ethyl 6-oxohexanoate ≥95%
Ethyl 6-oxohexanoate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.1950 g/mol
MDL Number: MFCD17170722
CAS Number: 27983-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR