3-Phenylpyridine-2-carboxylic acid ≥96%

Supplier: Ambeed
Warning

Synonyms: 3-Phenylpicolinic acid

A368724-1G A368724-250MG
77616-618EA 196.73 USD
77616-618 77616-620
3-Phenylpyridine-2-carboxylic acid ≥96%
3-Phenylpyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
MDL Number: MFCD04114112
CAS Number: 103863-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR