Methyl 2-(2-chloropyridin-3-yl)acetate ≥97%

Supplier: Ambeed
Warning

A368579-1G A368579-250MG
77616-616EA 63.86 USD
77616-616 77746-740
Methyl 2-(2-chloropyridin-3-yl)acetate ≥97%
Methyl 2-(2-chloropyridin-3-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
CAS Number: 123222-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR