N-Lauroyl-L-alanine ≥97%

Supplier: Ambeed
Warning

Synonyms: N-Lauroyl-Ala-OH

A368242-25G A368242-5G
77668-700EA 61 USD
77668-700 77668-702
N-Lauroyl-L-alanine ≥97%
N-Lauroyl-L-alanine

Precaución: for Research Use Only

Formula: C₁₅H₂₉NO₃
MW: 271.39 g/mol
MDL Number: MFCD00216726
CAS Number: 52558-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR