2-(4-((tert-Butoxycarbonyl)amino)cyclohexyl)acetic acid ≥97%

Supplier: Ambeed
Warning

A374559-100MG A374559-1G A374559-250MG A374559-5G
77626-730EA 62.83 USD
77626-730 77626-732 77626-734 77626-736
2-(4-((tert-Butoxycarbonyl)amino)cyclohexyl)acetic acid ≥97%
2-(4-((tert-Butoxycarbonyl)amino)cyclohexyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MDL Number: MFCD11848120
CAS Number: 344933-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR