((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoyl)glycine ≥97%

Supplier: Ambeed
Warning

A374346-25G A374346-5G A374346-100G A374346-10G A374346-1G
77601-654EA 154.78 USD
77601-654 77601-656 77626-724 77626-726 77626-728
((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoyl)glycine ≥97%
((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoyl)glycine

Precaución: for Research Use Only

Formula: C₂₆H₄₃NO₆
MDL Number: MFCD00065902
CAS Number: 475-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR