7-Methyl-1H-indazole-5-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (7-Methyl-1H-indazol-5-yl)boronic acid

A374181-100MG A374181-1G A374181-250MG
77885-986EA 36 USD
77885-986 77885-987 77885-988
7-Methyl-1H-indazole-5-boronic acid ≥98%
7-Methyl-1H-indazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MDL Number: MFCD12028646
CAS Number: 1245816-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR