cis-2-(tert-Butoxycarbonyl)octahydrocyclopenta[c]pyrrole-5-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A373582-100MG A373582-1G A373582-250MG
77890-766EA 278 USD
77890-766 77890-767 77890-768
cis-2-(tert-Butoxycarbonyl)octahydrocyclopenta[c]pyrrole-5-carboxylic acid ≥98%
cis-2-(tert-Butoxycarbonyl)octahydrocyclopenta[c]pyrrole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₄
MDL Number: MFCD28396323
CAS Number: 1069113-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR