2-Phenylethyl isothiocyanate ≥98%

Supplier: Ambeed
Danger

Synonyms: β-Phenethyl isothiocyanate, 2-Phenethyl isothiocyanate, 2-Isothiocyanatoethylbenzene, Phenethyl isothiocyanate

A367709-5G A367709-1G
77616-606EA 30.9 USD
77616-606 77748-732
2-Phenylethyl isothiocyanate ≥98%
2-Phenylethyl isothiocyanate

Precaución: for Research Use Only

Formula: C₆H₅CH₂CH₂NCS
MDL Number: MFCD00004821
CAS Number: 2257-09-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR