2,6-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester ≥97%

Supplier: Ambeed
Warning

A367685-100MG A367685-250MG
77872-564EA 126 USD
77872-564 77872-565
2,6-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester ≥97%
2,6-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD11053736
CAS Number: 1086394-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR