5-(N-Ethyl-N-isopropyl)amiloride ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-N-carbamimidoyl-6-chloro-5-(ethyl(isopropyl)amino)pyrazine-2-carboxamide, EIPA
A367526-10MG
A367526-1MG
A367526-5MG
77598-454EA
69.7
USD
77598-454
77598-456
77598-458
5-(N-Ethyl-N-isopropyl)amiloride ≥98%
5-(N-Ethyl-N-isopropyl)amiloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₈ClN₇O MW: 299.763 g/mol |
MDL Number:
MFCD00151710 CAS Number: 1154-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...