1-Nitro-2,3,4,5-tetrafluorobenzene ≥99%

Supplier: Ambeed
Warning

Synonyms: 2,3,4,5-Tetrafluoronitrobenzene, 1,2,3,4-Tetrafluoro-5-nitrobenzene

A373256-25G A373256-5G
77616-642EA 80.34 USD
77616-642 77616-644
1-Nitro-2,3,4,5-tetrafluorobenzene ≥99%
1-Nitro-2,3,4,5-tetrafluorobenzene

Precaución: for Research Use Only

Formula: C₆HF₄NO₂
MW: 195.07 g/mol
MDL Number: MFCD00014686
CAS Number: 5580-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR