1-Isopropylpiperidin-3-ol ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A367435-1G A367435-5G
77684-068EA 10 USD
77684-068 77892-753
1-Isopropylpiperidin-3-ol ≥98%, Ambeed.Inc
1-Isopropylpiperidin-3-ol

Precaución: For research use only.

Formula: C₈H₁₇NO
MDL Number: MFCD13190409
CAS Number: 3554-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR