2-Fluoro-4-(trifluoromethoxy)aniline ≥98%

Supplier: Ambeed
Danger

A367399-250MG A367399-5G
77892-751EA 61 USD
77892-751 77892-752
2-Fluoro-4-(trifluoromethoxy)aniline ≥98%
2-Fluoro-4-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₄NO
MDL Number: MFCD15527448
CAS Number: 123572-58-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR